EMAIL THIS PAGE TO A FRIEND
The American journal of tropical medicine and hygiene
Hookworm infection among school age children in Kintampo north municipality, Ghana: nutritional risk factors and response to albendazole treatment.
PMID 23836564
Abstract
Children (n = 812) 6-11 years of age attending 16 schools in the Kintampo North Municipality of Ghana were screened for participation in a study on hookworm infection, nutrition, and response to albendazole. The prevalence of Necator americanus hookworm infection (n = 286) was 39.1%, and significant predictors of infection included age, malaria parasitemia, lack of health care, school area, levels of antibodies against hookworm, and low consumption of animal foods. The cure rate after a single dose (400 mg) albendazole was 43%, and the mean fecal egg count reduction rate was 87.3%. Data for an in vitro egg hatch assay showed a trend toward reduced albendazole susceptibility in post-treatment hookworm isolates (P = 0.06). In summary, hookworm infection is prevalent among school age children in the Kintampo North Municipality and animal food intake inversely correlates with infection status. Modest cure rates and fecal egg count reduction rates reinforce the need for further investigation of potential benzimidazole resistance in Ghana.