EMAIL THIS PAGE TO A FRIEND
Nanotoxicology
Using gold nanorods core/silver shell nanostructures as model material to probe biodistribution and toxic effects of silver nanoparticles in mice.
PMID 23837638
Abstract
The aim of this work was to probe the biodistribution and toxic effects of silver nanoparticles (NPs) with powerful anti-bacterial and anti-virus activities. For this purpose, novel silver NPs with gold nanorod (NR) core and silver shell (Au@Ag NRs) were developed and employed as a model material. The inner gold core provided an excellent internal reference for tracking the NRs in vivo. After subcutaneous injection of Au@Ag NRs, silver and gold contents in the subcutis and organs were examined by inductively coupled plasma mass spectrometry at different time points within 28 days. Histological analysis, physiological function and complement faction 3 (C3) and 5a (C5a) measurement were performed over time to reveal the toxic effect of Au@Ag NRs in vivo. Experimental results showed that majority of the Au@Ag NRs remained in the injection site except for a small amount migrating into the lymph nodes. The silver shell was dissolved in the subcutaneous tissue and released silver ions rapidly, which resulted in detectable silver accumulation in most of the organs. The accumulated silver ions in the kidney not only interacted with the kidney cells membrane but also induced a rapid increase of complement fraction C3 followed by a significant consumption and C3a and C5a production significantly in the serum, which resulted in kidney oxidative damage and eventually led to the morphological changes and filtration function impairment of the glomerulus. The released silver ions also caused oxidative injury of subcutaneous tissue in the injection site.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
326542
Gold, beads, 1-6 mm, 99.999% trace metals basis
Au
373168
Gold, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Au
373184
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Au
265829
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Au
349240
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Au
326496
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Au
265810
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Au
349275
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Au
268461
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Au
349267
Gold, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Au
255734
Gold, powder, <850 μm, ≥99.99% trace metals basis
Au
265772
Gold, powder, <45 μm, 99.99% trace metals basis
Au
326585
Gold, powder, <10 μm, ≥99.9% trace metals basis
Au
636347
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Au
265837
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Au
265802
Gold, wire, diam. 2.0 mm, 99.99% trace metals basis
Au
349291
Gold, wire, diam. 1.5 mm, 99.999% trace metals basis
Au
349305
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Au
265799
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Au
349313
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Au
310980
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Au
265780
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Au
326534
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Au
326526
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Au
349283
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Au
GF43139538
Gold, insulated wire, 5m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Au
GF92307922
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyester insulation, 99.99%
Au
GF11444611
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation, 99.99%
Au
GF80583701
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.009mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Au
GF06766713
Gold, insulated wire, 5m, conductor diameter 0.125mm, insulation thickness 0.014mm, polyester insulation, 99.99%
Au