EMAIL THIS PAGE TO A FRIEND
Chemico-biological interactions
Effects of succinylcholine in an organotypic spinal cord-skeletal muscle coculture of embryonic mice.
PMID 23838346
Abstract
Intoxication with organophosphorus compounds is an important clinical problem worldwide. Although the core treatments - atropine, oximes and diazepam - are defined, high case fatalities were reported for intoxication with organophosphorus insecticides. In particular the role of oximes is not completely understood since they might benefit only patients poisoned by specific pesticides or patients with moderate poisoning and few randomised trials of such poisoning have been performed. This justifies the need for new in vitro test-systems like cocultures of spinal cord and muscle tissue, which have been recently introduced. However this test-system is not yet fully characterized. In order to estimate the applicability of cocultures of spinal cord and muscle tissue their sensitivity to succinylcholine (di-acetylcholine), a depolarizing muscle relaxant in clinical use, was tested. The test-system evaluated in the current study showed sensitivity to succinylcholine with an EC50 as low as 1μM thereby being close to the EC50 value in adult human patients (2.6μM). Furthermore, action potential activity of spinal ventral horn neurons was not altered by succinylcholine. The latter observations strongly suggest that our preparation well predicts the qualitative and quantitative actions of novel drugs targeting the neuromuscular system in vivo. In summary, cocultures of spinal cord and muscle tissue seem to be a valid test-system for the development and investigation of new oximes. Moreover, practical aspects like transport over long distances to further laboratories, the opportunity to conduct long-term studies and the reduction of animal usage display further advantages of its use.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8251
Succinylcholine chloride dihydrate, 98.0-102.0%, solid
C14H30Cl2N2O4 · 2H2O