EMAIL THIS PAGE TO A FRIEND
European journal of mass spectrometry (Chichester, England)
Letter: Mass spectrometric approach of high pH- and copper-induced glutathione oxidation.
PMID 23841227
Abstract
The interaction between copper ions and gamma-L-glutamyl-L-cysteinyl-glycine [glutathione (GSH)] molecules may lead to the formation of the physiologically occurring Cu[I)-[GSH]2 and Cu(II)-GSSG complexes. Since glutathione depletion in neurons and aberrant copper metabolism have been implicated in several neurodegenerative disorders, we studied here the interaction of GSH with copper ions (Cu2+) by electrospray ionization ion trap mass spectrometry (ESI-IT-MS). Besides, we extended this approach to pH in excess of 10 by adding ethanolamine to the solution being investigated. As a result, the ESI-IT-MS spectra revealed novel aspects regarding the speciation of copper-glutathione complex.