EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong xi yi jie he za zhi Zhongguo Zhongxiyi jiehe zazhi = Chinese journal of integrated traditional and Western medicine
[Treatment of refractory depression patients of yin deficiency inner heat syndrome by jieyu granule combined paroxetine: an efficacy observation].
PMID 23841263
Abstract
To explore the efficacy of jieyu granule (JG) combined Paroxetine in treating refractory depression (RD) patients of yin deficiency inner heat syndrome (YDIHS). Seventy RD patients of YDIHS were randomly assigned to the experimental group (JG combined Paroxetine) and the control group (Chinese medical placebo combined Paroxetine), 35 cases in each group. Hamilt Depression Rating Scale and Hamilton Anxiety Scale were used before treatment, and at the weekend of the 2nd, 4th, and 8th week, respectively. In the experimental group, 32 patients completed the trial and 3 patients dropped out. In the control group, 33 patients completed the trial and 2 patients dropped out. At the end of the 8th week of the treatment, the total score of Hamilt Depression Rating Scale was (14.75 +/- 7.85) in the experimental group, lower than that of the control group (19. 06 +/- 8. 31, P <0.05). At the end of the 2nd, 4th, and 8th week of the treatment, the score of Hamilton Anxiety Scale was 17.03 +/- 4.25, 14.50 +/- 5. 13, and 11.03 +/- 4.88, respectively in the experimental group, lower than that of the control group at each corresponding time point (19. 60 +/-3. 96, 17. 12 +/- 4.14, 14.64 +/- 4.47, P <0.05, P <0.01). The efficacy of JG combined Paroxetine for treating RD patients of YDIHS was superior to that of using Paroxetine alone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000578 Paroxetine hydrochloride (anhydrous), European Pharmacopoeia (EP) Reference Standard
C19H20FNO3 · HCl