EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Case records of the Massachusetts General Hospital. Case 23-2013. A 54-year-old woman with abdominal pain, vomiting, and confusion.
PMID 23841704