EMAIL THIS PAGE TO A FRIEND
Nihon yakurigaku zasshi. Folia pharmacologica Japonica
Pharmacological and clinical properties of AIMIX® combination tablets LD&HD, fixed-dose combination of irbesartan and amlodipine besilate.
PMID 23842227