EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Ion-paired high-performance liquid chromatographic separation of trimethoprim, sulfamethoxazole and N4-acetylsulfamethoxazole with solid-phase extraction.
PMID 2384558
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000412
Sulfamethoxazole impurity A, European Pharmacopoeia (EP) Reference Standard
C12H13N3O4S