EMAIL THIS PAGE TO A FRIEND
Journal of Chromatographic Science
Development of stability-indicating UHPLC method for the quantitative determination of silodosin and its related substances.
PMID 23845884