EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong Yao Za Zhi = Zhongguo zhongYao ZaZhi = China Journal of Chinese Materia Medica
[HPLC fingerprint of ethyl acetate extraction of Saxifraga stolonifera].
PMID 23847951