EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Effects of sapindus saponins on inflammatory response mediated by Ang II/p38MAPK pathway and cardiac hypertrophy in spontaneously hypertensive rats].
PMID 23847952
Abstract
To investigate the effects of sapindus saponins on myocardial inflammation mediated by Ang II/ p38MAPK signal pathway and cardiac hypertrophy in spontaneously hypertensive rats. And also to explore the correlation of cardiac hypertrophy and inflammation. Thirty-two 16-week-old spontaneously hypertensive rats (SHR) were randomly divided into four groups, one with placebo as model group, one with captopril tablets (27 mg x kg(-1)) as positive control, one with low-dose sapindus saponins (27 mg x kg(-1)), one with high-dose (108 mg x kg(-1)). And another eight healthy Wistar-Kyoto strain (WKY) rats were used as the normal group. The animals were treated for eight weeks, and the indicators detected were as follows: (1) left ventricular mass index (LVMI); (2) the content of Ang II and hs-CRP in plasma were determined by ELISA; (3) the protein expression of AT1R and VEGF were determined by immunohistochemical method; (4) the protein expression of p-p38MAPK in myocardial cells was determined by Western blot. Sapindus saponins reduced LVMI, and blocked the expression level of Ang II, AT1R, p-p38MAPK, VEGF and hs-CRP in myocardial tissue. Vs the SHR model group, there were significant differences (P < 0.05 or P < 0.01). Our findings suggested that sapindus saponins could inhibited cardiac hypertrophy, the possible mechanisms may be related to the inhibition on inflammatory response mediated by Ang II/p38MAPK pathway.