EMAIL THIS PAGE TO A FRIEND
Lancet (London, England)
SGLT2 inhibitors for diabetes: turning symptoms into therapy.
PMID 23850056
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000516
Glimepiride for system suitability, European Pharmacopoeia (EP) Reference Standard
C24H34N4O5S