EMAIL THIS PAGE TO A FRIEND
The Journal of steroid biochemistry and molecular biology
Re: effect of finasteride on serum levels of androstenedione, testosterone and their 5α-reduced metabolites in men at risk for prostate cancer.
PMID 23851167