EMAIL THIS PAGE TO A FRIEND
Canadian family physician Medecin de famille canadien
Anaphylaxislike cholinergic urticaria.
PMID 23851539
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001434 Montelukast sodium, European Pharmacopoeia (EP) Reference Standard
C35H35ClNO3S·Na