EMAIL THIS PAGE TO A FRIEND
Organic & Biomolecular Chemistry
Chemoselective alcoholysis/acetolysis of trans-ketals over cis-ketals and its application in the total synthesis of the cellular second messenger, D-myo-inositol-1,4,5-trisphosphate.
PMID 23851711