EMAIL THIS PAGE TO A FRIEND
The Journal of clinical pediatric dentistry
Deproteinization treatment on bond strengths of primary, mature and immature permanent tooth enamel.
PMID 23855172
Abstract
The aim of this study was to examine the effect of pre-post deproteinization treatment with 5% sodium hypochloride on shear bond strength (sbs) of adhesive resin to primary, immature and mature permanent teeth enamel. 30 teeth were used for each of primary, immature and mature permanent teeth groups. (totally 90). In control groups, enamel was etched for 60s with 37% phosphoric acid (3M) and rinsed for 10s (Procedure A). In experimental groups, deproteinization was applied with 5% NaOCI solution for 120s before (Procedure D+A) and after acid-etching (Procedure A+D). Gluma Comfort Bond (Heraeus-Kulzer) and Charisma (Heraeus-Kulzer) composite resin were applied to etched enamel surfaces. Data were determined with Two-Way ANOVA and LSD Multiple Comparison Test (p < 0.05). SBS was significantly lower in primary and immature permanent teeth than mature permanent teeth (p < 0.05). "Procedure A+D" statistically increased sbs values in primary and immature permanent teeth (p < 0.05). Deproteinization after acid etching significantly enhanced the shear bond strength values in primary and immature permanent teeth.