EMAIL THIS PAGE TO A FRIEND
Nippon Ganka Gakkai zasshi
[Influence on ocular surface and intraocular pressure of switching from preservative-containing to preservative-free carteolol hydrochloride ophthalmic solution].
PMID 23855234
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000193 Carteolol for system suitability, European Pharmacopoeia (EP) Reference Standard
C16H24N2O3 · HCl