EMAIL THIS PAGE TO A FRIEND
Chirurgie de la main
[Treatment of Dupuytren's contracture by collagenase injection].
PMID 23856551
Abstract
Dupuytren's disease is a disorder of the palmar aponeurosis responsible for apparition of pathological collagen cords that will gradually lead to a digital retraction. These cords cause functional disability to the hand with a real handicap for the patient. No curative treatment exists currently in this disease. Injections of collagenase from Clostridium histolyticum cause lysis of the collagen present in the cords and cause an interruption of the palmar cord. It is a new treatment in the management of Dupuytren's disease. The use of collagenase injection in the treatment of palmar form of Dupuytren's disease gives good results in the short and medium terms. It is a simple, minimally invasive treatment, several studies have proven its effectiveness and it approximates percutaneous needle fasciotomy in its easy use and its indications. Patients should be well informed about local reactions and transient pain with the injection of C. histolyticum collagenase. Diffusion of this technique is still limited by: the assessment of its cost compared to that of other methods of treatment and particularly the needle fasciotomy, and the evaluation of results over the long-term.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6079 Collagenase + protease inhibitor, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid, Suitable for isolation of rat pancreatic islet cells.
C1889
Collagenase from Clostridium histolyticum, sterile-filtered, release of physiologically active rat hepatocytes tested, Type IV-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
C0773
Collagenase from Clostridium histolyticum, high purity, purified by chromatography, Type VII, ≥4 FALGPA units/mg solid, lyophilized powder, ≥700 CDU/mg solid (CDU = collagen digestion units)
C1639
Collagenase from Clostridium histolyticum, sterile-filtered, for general use, Type I-S, 0.2-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid
C1764
Collagenase from Clostridium histolyticum, sterile-filtered, release of physiologically active rat epididymal adipocytes tested, Type II-S, 0.5-5.0 FALGPA units/mg solid
C9722
Collagenase from Clostridium histolyticum, lyophilized powder (from sterile-filtered solution), 0.5-5.0 FALGPA units/mg solid, suitable for cell culture
C5894
Collagenase from Clostridium histolyticum, sterile; sterile-filtered, Type IA-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
C9697
Collagenase from Clostridium histolyticum, lyophilized powder (from sterile-filtered solution), Suitable for digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture
C7926
Collagenase from Clostridium histolyticum, Sigma Blend Type F, ≥2.0 FALGPA units/mg solid
C2399
Collagenase from Clostridium histolyticum, sterile-filtered, high purity, purified by chromatography, Type VII-S, ≥4 FALGPA units/mg solid, ≥700 CDU/mg solid (CDU = collagen digestion units)
C0255
Collagenase from Clostridium histolyticum, purified by chromatography, ≥0.25 FALGPA units/mg solid, ≥250 CDU/mg solid (CDU = collagen digestion units), lyophilized powder
C2014
Collagenase from Clostridium histolyticum, sterile-filtered, suitable for release of physiologically active rat pancreatic islets, Type V-S, ≥1 FALGPA units/mg solid, ≥125 CDU/mg solid
C2799
Collagenase from Clostridium histolyticum, powder, suitable for cell culture, ≥4 FALGPA units/mg solid, high purity, ≥700 CDU/mg solid (CDU = collagen digestion units)
C8176
Collagenase from Clostridium histolyticum, Sigma Blend Type L, ≤1.0 FALGPA units/mg solid