EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
Synthesis of copper nanoparticles with controlled sizes by reverse micelle method.
PMID 23858839
Abstract
Copper (Cu) nanoparticles with controllable sizes were successfully synthesized by using a modified reverse micelle method, in which copper(II) acetate was reacted with L-ascorbic acid in a solution containing water and xylene in the presence of oleic acid and oleylamine as surfactants. The as-synthesized Cu nanoparticles had a nearly spherical profile and multiple-twinned structures, whose surface was enclosed by {111} facets. Our synthetic procedure provides a simple and readily scalable route to the preparation of Cu nanoparticles with controllable sizes.