EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
Synthesis and electrochemical characterization of LiMn0.6Fe0.4PO4/C cathode material via a modified-solid state reaction method.
PMID 23858843
Abstract
LiMn0.6Fe0.4PO4/C cathode material is synthesized via a modified-solid state reaction method. The calcination temperature is adjusted in the range of 500-700 degrees C for 10 h. The crystal structure, morphology, and carbon coating layer of the synthesized LiMn0.6Fe0.4PO4/C are analyzed using X-ray diffraction (XRD), scanning electron microscopy (SEM), and transmission electron microscopy (TEM), respectively. The electrochemical performance of LiMn0.6Fe0.4PO4/C, such as initial capacity, rate capability, cycling performance and EIS is also evaluated. The synthesized cathode material shows around 100-200 nm of primary particle size with no impurities. The highest initial discharge capacity of 162.1 mA h g(-1) and columbic efficiency of 98.5% are obtained at a heat treatment temperature of 600 degrees C. In addition, LiMn0.6Fe0.4PO4/C active material shows the high capacity retention of 85% at 5 C compared to 0.2 C. It also shows the excellent capacity retention of 97.5% after the 50th charge/discharge.