EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
A new deposition method for the preparation of Pt-Pd nanocatalysts on a nafion coated carbon black via the reduction of metallic precursors in a drying process.
PMID 23858917
Abstract
A simple method for the synthesis of Pt-Pd nanocatalysts was developed for a proton-exchange membrane fuel cell (PEMFC), which was loaded on a nafion coated carbon black via the sequential reduction of palladium(II) bis(acetylacetonato) and platinum(II) bis(acetylacetonato) in a drying process. Metallic precursors were sublimed and reduced on a nafion coated carbon black which was spray coated on a gas diffusion layer (GDL) in a glass reactor of N2 atmosphere at 180 degrees C for various times. The morphology and distribution of the Pt and Pd nanoparticles were observed by scanning electron microscopy (SEM) and transmission electron microscopy (TEM), and we found that the loading weight, number density and particle size of Pt-Pd nanoparticles increased with increasing exposure time at 180 degrees C.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
326666
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Pd
686468
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Pd
203939
Palladium, powder, 99.995% trace metals basis
Pd
464651
Palladium, powder, <75 μm, 99.9% trace metals basis
Pd
203998
Palladium, granular, 99.99% trace metals basis
Pd
267082
Palladium, sponge, 99.9% trace metals basis
Pd
267112
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Pd
373206
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Pd
348643
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Pd
287474
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Pd
348694
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Pd
267120
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Pd
348678
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Pd
411450
Palladium, foil, thickness 0.05 mm, 99.9% trace metals basis
Pd
373192
Palladium, evaporation slug, diam. × L 0.9 cm × 1.2 cm, 99.95% trace metals basis
Pd
346985
Palladium, rod, diam. 6.25 mm, ≥99.9% trace metals basis
Pd
643181
Palladium, extent of labeling: 5 wt. % loading (dry basis), matrix activated carbon, wet support, Degussa type E105CA/W
Pd
520810
Palladium black, surface area 40-60 m2/g, 99.95% trace metals basis
Pd
267228
Platinum, wire, diam. 0.5 mm, 99.9% trace metals basis
Pt
267201
Platinum, wire, diam. 0.5 mm, 99.99% trace metals basis
Pt
298093
Platinum, gauze, 100 mesh, 99.9% trace metals basis
Pt
327492
Platinum, wire, diam. 1.0 mm, 99.9% trace metals basis
Pt
267244
Platinum, foil, thickness 0.025 mm, 99.9% trace metals basis
Pt
298107
Platinum, gauze, 52 mesh, 99.9% trace metals basis
Pt
267236
Platinum, wire, diam. 1.0 mm, 99.99% trace metals basis
Pt
267163
Platinum, wire, diam. 0.127 mm, 99.99% trace metals basis
Pt
349356
Platinum, foil, thickness 0.05 mm, 99.99% trace metals basis
Pt
267260
Platinum, foil, thickness 0.5 mm, 99.99% trace metals basis
Pt
685453
Platinum, nanopowder, <50 nm particle size (TEM)
Pt
357359
Platinum, wire, diam. 0.20 mm, ≥99.99% trace metals basis
Pt