EMAIL THIS PAGE TO A FRIEND
Journal of biomedical nanotechnology
Engineering bioconjugated gold nanospheres and gold nanorods as label-free plasmon scattering probes for ultrasensitive multiplex dark-field imaging of cancer cells.
PMID 23858962
Abstract
In this paper, we proposed the use of gold nanoparticles as plasmon scattering probes for dark-field multiplex imaging of live cancer cells. By carefully engineering the surfaces, aqueous dispersions of anti-EGFR antibody-conjugated gold nanospheres and gold nanorods are prepared. We demonstrated the receptor-mediated delivery of antibody conjugated gold nanospheres and gold nanorods into EGFR receptor-positive oral squamous cell carcinoma cell line by using darkfield microscopy technique. Our result suggests that gold nanospheres and gold nanorods formulations could provide up to 7 types of plasmon scattering probes based on their tunability range with a 100 nm spectral separation in absorption bands.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
326542
Gold, beads, 1-6 mm, 99.999% trace metals basis
Au
373168
Gold, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Au
373184
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Au
265829
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Au
349240
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Au
326496
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Au
265810
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Au
349275
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Au
268461
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Au
349267
Gold, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Au
255734
Gold, powder, <850 μm, ≥99.99% trace metals basis
Au
265772
Gold, powder, <45 μm, 99.99% trace metals basis
Au
326585
Gold, powder, <10 μm, ≥99.9% trace metals basis
Au
636347
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Au
265837
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Au
265802
Gold, wire, diam. 2.0 mm, 99.99% trace metals basis
Au
349291
Gold, wire, diam. 1.5 mm, 99.999% trace metals basis
Au
349305
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Au
265799
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Au
349313
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Au
310980
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Au
265780
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Au
326534
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Au
326526
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Au
349283
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Au
643254
Gold coated glass cover slip, 99.999% (Au), layer thickness 100 Å, L × W × thickness 22 mm × 22 mm × 130-170 μm, square
Au
643289
Gold coated glass cover slip, layer thickness 100 Å, diam. × thickness 15 mm × 130-170 μm, 99.999% (Au)
Au
643246
Gold coated microscope slide, layer thickness 1000 Å, 99.999% (Au)
Au
643203
Gold coated microscope slide, layer thickness 100 Å, 99.999% (Au)
Au