EMAIL THIS PAGE TO A FRIEND
Molecular carcinogenesis
Inhibitions of epithelial to mesenchymal transition and cancer stem cells-like properties are involved in miR-148a-mediated anti-metastasis of hepatocellular carcinoma.
PMID 23861222
Abstract
The epithelial-mesenchymal transition (EMT) and acquisition of cancer stem cells (CSCs)-like properties are essential steps in the metastasis and postsurgical recurrence of hepatocellular carcinomas (HCCs). The molecular mechanisms involved, however, remain obscure. As determined by an miRNA microarray analysis, there was lower expression of miR-148a in poorly differentiated HCC tissues relative to well-differentiated HCC tissues. MHCC97H and MHCC97L (HCC cells with migratory capacity) and HCC tissues with various differentiation status were selected for further investigation. The results showed that miR-148a levels inversely correlated with the differentiation status of HCC tissues. In MHCC97H and MHCC97L cells, over-expression of miR-148a blocked the EMT process, attenuated the expression of CD90 and CD44 (biomarkers for liver cancer stem cells), and inhibited their migratory capacity. Via TargetScan and microRNA.org algorithms, miR-148a was predicted to bind to the Wnt1 mRNA 3'-UTR. Wnt1 was confirmed as a target gene of miR-148a in HCC cells, and the Wnt signal pathway was determined to be involved in the miR-148a-mediated inhibition of EMT and CSCs-like properties of MHCC97H cells. Moreover, the expression of miR-148a in nonmetastatic HCC tissues was higher than that in metastatic HCC tissues. The results suggest that miR-148a inhibits the metastasis of HCCs by blocking EMT and CSCs-like properties through effects on the Wnt signaling pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21186 Anti-CDH1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501665
Anti-CDH1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2701861
Anti-cdh1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA004812
Anti-CDH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107370
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB4300693
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB2104222
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
C7855
Anti-Cdh1 antibody,Mouse monoclonal, clone DCS-266, purified from hybridoma cell culture
EMU024721
MISSION® esiRNA, targeting mouse Wnt1
SAB4700237 Monoclonal Anti-CDH1 antibody produced in mouse, clone 67A4, purified immunoglobulin, buffered aqueous solution
WH0000999M1
Monoclonal Anti-CDH1 antibody produced in mouse, clone 3F4, purified immunoglobulin, buffered aqueous solution
AMAB90862
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1170, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90863
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1172, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90865
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1180, purified immunoglobulin, buffered aqueous glycerol solution