EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
Calcium ions promote superoxide dismutase 1 (SOD1) aggregation into non fibrillar amyloid: a link to toxic effects of calcium overload in amyotrophic lateral sclerosis (ALS)?
PMID 23861388