EMAIL THIS PAGE TO A FRIEND
Journal of central nervous system disease
Sertindole in the management of schizophrenia.
PMID 23861640
Abstract
Nowadays, new schizophrenia treatments are more ambitious than ever, aiming not only to improve psychotic symptoms, but also quality of life and social reinsertion. Our objective is to briefly but critically review the diagnosis of schizophrenia, the atypical antipsychotics sertindole's pharmacology, safety and status, and mainly evaluate the effects of sertindole compared with other second generation antipsychotics for people with schizophrenia and schizophrenia-like psychosis. In vitro studies showed that sertindole exerts a potent antagonism at serotonin 5-HT2A, 5-HT2C, dopamine D2, and αl adrenergic receptors. Sertindole offers an alternative treatment option for refractory patients given its good EPS profile, favorable metabolic profile, and comparable efficacy to risperidone. Due to cardiovascular safety concerns, sertindole is available as a second-line choice for patients intolerant to other antipsychotic agents. Further clinical studies, mainly comparisons with other second-generation antipsychotic agents, are needed to define the role of sertindole in the treatment of schizophrenia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8072
Sertindole, ≥97.5% (HPLC)
C24H26ClFN4O