EMAIL THIS PAGE TO A FRIEND
Rossiiskii fiziologicheskii zhurnal imeni I.M. Sechenova
[Features of spatial orientation formation in posterity of female rats with liver experimental defeat in Morris water maze].
PMID 23862386
Abstract
It was established, that the chronic experimental hepatobiliary systems alcohol induced pathology of female rats caused infringement of spatial differentiation development in their posterity in Morris water maze. In experimental infant rats it was revealed the increase of time of platform fin ding in a water Morris task, lengths of trajectory of movement, increase of time of stay near walls (thigmotaxis) and speed of movement reduction, the latent period of zones of start leaving and efficiency of way index.