EMAIL THIS PAGE TO A FRIEND
Gan to kagaku ryoho. Cancer & chemotherapy
[A case of necrotizing fasciitis developed in a patient with recurrent rectal cancer treated with chemotherapy].
PMID 23863596
Abstract
A fifty-two-year-old female with advanced rectal cancer underwent low anterior resection and hysterectomy. After local pelvic recurrence, she underwent radiation therapy(50 Gy in total), and then chemotherapy(XELOX+bevacizumab)was started. However, left femoral pain developed after the second course of chemotherapy, and necrotizing fasciitis associated with lower bowel perforation was found. Although bowel perforation is known as a serious side-effect of bevacizumab, its association with fasciitis is rare.