EMAIL THIS PAGE TO A FRIEND
Gan to kagaku ryoho. Cancer & chemotherapy
[A retrospective study of UFT and oral leucovorin combination adjuvant chemotherapy for patients over 76 years old with stage III colon cancer].
PMID 23863728
Abstract
To perform a retrospective analysis of UFT and oral leucovorin combination adjuvant chemotherapy for Stage III colon cancer patients over 76 years old, in order to evaluate both treatment efficacy and toxicity. Between 2002 and 2011, 333 Stage III colon cancer patients had surgery performed in our institute, and we studied 25 of them on our chemotherapy regimen. Patients'median age was 78 years old, with 12 men and 13 women. Of all the patients, 19 had Stage IIIa and 6 had Stage IIIb. The 3-year disease-free survival rates for Stage III and Stage IIIa patients were 65. 1% and 83. 1%, respectively, and the 3-year overall survival rate for Stage III was 79. 9%. With regard to toxicity, liver function disorder was observed in 8% of the patients, being the adverse event that occurred the most, but there was no Grade 3 or 4 toxicity. UFT and oral leucovorin combination adjuvant chemotherapy for Stage III colon cancer patients over 76 years showed a good response, especially for Stage III a.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47612
Folinic acid calcium salt hydrate, BioXtra, ≥99.0% (HPLC)
C20H21CaN7O7 · xH2O
F7878
Folinic acid calcium salt hydrate, analytical standard
C20H21CaN7O7 · xH2O