EMAIL THIS PAGE TO A FRIEND
Archives of environmental health
Aldicarb food poisonings in California, 1985-1988: toxicity estimates for humans.
PMID 2386419
Abstract
Three outbreaks of food poisoning involving watermelons or cucumbers and caused by the carbamate pesticide aldicarb occurred in California between 1985 and 1988. For each outbreak, and for an outbreak of aldicarb poisoning associated with English cucumbers previously reported in the literature, dosages of aldicarb sulfoxide that caused the illnesses were estimated. Estimated dosages ranged between 0.0023 [corrected] and 0.06 mg/kg body weight, and most were well below the 0.025 mg/kg Lowest Observed Effect Level (LOEL) for subclinical blood cholinesterase depression previously reported for humans. These findings are consistent with aldicarb sulfoxide (ASO) illnesses that have occurred in other states. Aldicarb appears to be more toxic than previously suspected. Scientific and regulatory implications are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31258
Aldicarb-sulfoxide, PESTANAL®, analytical standard
C7H14N2O3S