EMAIL THIS PAGE TO A FRIEND
Sheng wu yi xue gong cheng xue za zhi = Journal of biomedical engineering = Shengwu yixue gongchengxue zazhi
[Study on preparation and property of a new adsorbent for endotoxin removal in blood purification].
PMID 23865333
Abstract
In order to remove the endotoxin from the blood of endotoxemia patients, we prepared a new adsorbent with heparin space arm and polymyxin B (PMB) ligand. The carrier of chloromethyl polystyrene resin was activated and heparin space arm was grafted, and then PMB ligand was immobilized onto adsorbent with glutaraldehyde. We employed in vitro FITC-lipopolysaccharide (FITC-LPS) static adsorption to characterize the adsorption properties on the adsorbent, and conducted in vitro lipopolysaccharide (LPS) static adsorption to measure quantitavely the adsorption capacity and rate, and then evaluated the blood compatibility. The in vitro static adsorption indicated that the adsorbent had the removal rate of LPS above 70% with the adsorption equilibrium time for 2 hours. Blood compatibility experiment showed that the adsorbent had little negative effects on blood cells and plasma protein, and their adsorption rates were less than 10% for hemocytes and 20% for plasma protein respectively. This adsorbent exhibited high selectivity, high adsorption capacity and good biocompatibility, and presented a promising clinical application in the treatment of endotoxemia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F8666 Lipopolysaccharides from Escherichia coli O55:B5, FITC conjugate