EMAIL THIS PAGE TO A FRIEND
Technology Evaluation Center Assessment Program. Executive summary
Sequencing-based tests to determine fetal down syndrome (trisomy 21) from maternal plasma DNA.
PMID 23866392