EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Aqueous synthesis of glutathione-coated PbS quantum dots with tunable emission for non-invasive fluorescence imaging in the second near-infrared biological window (1000-1400 nm).
PMID 23867970
Abstract
Glutathione-coated PbS quantum dots with tunable emission were synthesized in the aqueous phase and used for non-invasive tissue imaging in the second near-infrared biological window.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
372595
Lead(II) sulfide, 99.9% trace metals basis
PbS