EMAIL THIS PAGE TO A FRIEND
Food chemistry
Whey protein isolate polydispersity affects enzymatic hydrolysis outcomes.
PMID 23870966
Abstract
The effects of heat-induced denaturation of whey protein isolate (WPI) on the enzymatic breakdown of α-La, caseinomacropeptide (CMP), β-Lg A and β-Lg B were observed as hydrolysis proceeded to a 5% degree of hydrolysis (DH) in both unheated and heat-treated (80 °C, 10 min) WPI dispersions (100 g L(-1)). Hydrolysis of denatured WPI favoured the generation of higher levels of free essential amino acids; lysine, phenylalanine and arginine compared to the unheated substrate. LC-MS/MS identified 23 distinct peptides which were identified in the denatured WPI hydrolysate - the majority of which were derived from β-Lg. The mapping of the detected regions in α-La, β-Lg, and CMP enabled specific cleavage points to be associated with certain serine endo-protease activities. The outcomes of the study emphasise how a combined approach of substrate heat pre-treatment and enzymology may be used to influence proteolysis with attendant opportunities for targeting unique peptide production and amino acid release.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L7252 Lactalbumin