EMAIL THIS PAGE TO A FRIEND
Food chemistry
Effect of zinc sulphate on gelling properties of phosphorylated protein isolate from yellow stripe trevally.
PMID 23871033
Abstract
Impacts of zinc sulphate (ZnSO4) (0-140 μmol/kg) on gel properties of yellow stripe trevally surimi added with sodium tripolyphosphate (STPP) (0.25% and 0.5%, w/w) and protein isolate phosphorylated with STPP at 0.25% and 0.5% (w/w) were studied. Gels from surimi added with 60 μmol ZnSO4/kg in the absence and presence of 0.5% STPP had the increases in breaking force and deformation by 20.9% and 33.3%, and 11.6% and 18.6%, respectively, compared with the control surimi gel (without additives). Gel of protein isolate phosphorylated with 0.5% STPP containing 100 μmol ZnSO4/kg had the increases in breaking force and deformation by 14.87% and 5.6%, respectively, compared with the gel from non-phosphorylated protein isolate at the same ZnSO4 level, suggesting that the phosphorylated protein isolate was more crosslinked by Zn(2+). The addition of ZnSO4 at the suitable level lowered the expressible moisture content, but increased whiteness of surimi or protein isolate gels (P<0.05). Non-covalent bonds, more likely salt bridge and ionic interactions, played a major role in cross-linking of proteins in both surimi and protein isolate added with ZnSO4, regardless of phosphates incorporated. Microstructure study revealed that a gel having highly interconnected and denser network with smaller voids was formed when protein isolate phosphorylated with 0.5% STPP was added with ZnSO4 at a level of 100 μmol/kg. Thus, gel with improved properties could be obtained from protein isolate from yellow stripe trevally phosphorylated with STPP in conjunction with addition of ZnSO4 at an appropriate level.