EMAIL THIS PAGE TO A FRIEND
Food chemistry
Use of allyl isothiocyanate and carbon nanotubes in an antimicrobial film to package shredded, cooked chicken meat.
PMID 23871073
Abstract
We developed antimicrobial packaging incorporated with allyl isothiocyanate (AIT) and carbon nanotube (CNT), and this packaging was used for shredded cooked chicken meat inoculated with Salmonella Choleraesuis. The following parameters were analysed during the 40 days of storage: microbial counts, colour characteristics and changes in the oxidation of the meat as well as changes in the mechanical properties of the film, the structure of the antimicrobial film and the diffusion of the antimicrobial agent into the food. The incorporation of AIT into the films increased the elongation at the break (E) value of the films and decreased the tensile strength (TS) value of the films. The CNT was important to retain the AIT which is a volatile substance in the film. The diffusion of the AIT from the film into the chicken reduced the microbial contamination, controlled oxidation and reduced the colour changes. Thus, these packages were effective for the 40 days of storage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36682
Allyl isothiocyanate, PESTANAL®, analytical standard
C4H5NS