EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Clostridium acidurici electron-bifurcating formate dehydrogenase.
PMID 23872566
Abstract
Cell extracts of uric acid-grown Clostridium acidurici catalyzed the coupled reduction of NAD(+) and ferredoxin with formate at a specific activity of 1.3 U/mg. The enzyme complex catalyzing the electron-bifurcating reaction was purified 130-fold and found to be composed of four subunits encoded by the gene cluster hylCBA-fdhF2.