EMAIL THIS PAGE TO A FRIEND
European journal of ophthalmology
Riboflavin/UVA photochemical therapy for severe infectious keratitis.
PMID 23873492
Abstract
To describe the antibacterial activity of treatment with riboflavin and ultraviolet A light (UVA) in cases of severe infectious keratitis. A retrospective analysis was performed of an interventional case series in which 6 eyes of 6 patients with severe infectious keratitis, all of whom were refractive to multidrug conventional therapy, were treated with riboflavin/UVA. The procedure was conducted according to the standardized protocol of corneal collagen crosslinking (CXL) for keratoconus. Best spectacle-corrected visual acuity and clinical outcomes were evaluated before and during the follow-up period. Five of the 6 patients showed rapid reduction in symptoms and decreased infiltrate size after riboflavin/UVA photochemical therapy. Signs of infection and inflammation mostly resolved within 1 to 2 weeks after the treatment. Despite this therapy, one patient continued to deteriorate, and penetrating keratoplasty was performed. The adjunctive use of riboflavin/UVA photochemical therapy has a positive effect on refractory infectious keratitis. The treatment seems to be safe and effective and should be considered as part of the first-line therapy in severe cases of infectious keratitis.