EMAIL THIS PAGE TO A FRIEND
Poultry science
Linking ileal digestible phosphorus and bone mineralization in broiler chickens fed diets supplemented with phytase and highly soluble calcium.
PMID 23873559
Abstract
The objectives of this study were to determine the ileal digestibility of P in potassium phosphate, phytase-related ileal digestible P release, bone-mineralization-based ileal digestible P equivalency of phytase, and phytase-related efficiency of ileal digestible P utilization for bone mineralization in broiler chickens at 2 dietary concentrations of highly soluble Ca (HSC). Birds were sorted by BW at d 15 posthatch and assigned to 8 cages per diet with 8 birds per cage. Twelve diets were arranged in a 2 × 6 factorial of HSC at 5 or 6 g/kg and P supply treatment at 6 levels consisting of 4 added P levels (P from KH2PO4 added at 0, 0.7, 1.4, or 2.1 g/kg of diet) or 2 added phytase levels (500 or 1,000 phytase units). On d 24 posthatch, ileal digesta were collected for ileal P digestibility (IPD) determination and the left tibia was collected from the 4 heaviest birds in each cage for bone ash determination. Weight gain, G:F, and tibia ash were higher (P < 0.05) at 5 than at 6 g of HSC/kg. Added P from KH2PO4 or added phytase linearly increased (P < 0.001) weight gain, G:F, tibia ash, and IPD. The IPD of KH2PO4 derived from multiple linear regressions of digestible on total P intake for the diets without added phytase showed a reduction (P < 0.05) from 89.5 to 84.5% with increased HSC from 5 to 6 g/kg. Polynomial regressions of digestible P intake on phytase intake indicated that 1,000 units of added phytase released 1.701 or 1.561 g of digestible P in diets containing 5 or 6 g of added HSC/kg, respectively. Polynomial regressions of tibia ash on digestible P or phytase intake in diets containing 5 or 6 g of added HSC/kg at 1,000 phytase units gave digestible P equivalency of 1.487 or 1.448 g, respectively. Thus, phytase-related efficiency of ileal digestible P utilization for bone mineralization was 87.4 and 92.8% in diets containing 5 or 6 g of added HSC/kg, respectively.