PloS one
Overexpressed microRNA-182 promotes proliferation and invasion in prostate cancer PC-3 cells by down-regulating N-myc downstream regulated gene 1 (NDRG1).
PMID 23874837
Abstract
MicroRNAs, non-coding 20-22 nucleotide single-stranded RNAs, result in translational repression or degradation and gene silencing of their target genes, and significantly contribute to the regulation of gene expression. In the current study, we report that miR-182 expression was significantly upregulated in prostate cancer tissues and four cell lines, compared to benign prostatic hyperplasia tissues and normal prostatic epithelial (RWPE-1) cells. Ectopic overexpression of miR-182 significantly promotes the proliferation, increases the invasion, promotes the G1/S cell cycle transition and reduces early apotosis of PC-3 cells, while suppression of miR-182 decreased the proliferation and invasion, inhibits the G1/S cell cycle transition and increase early apotosis of PC-3 cells. Additionally, we demonstrated that miR-182 could downregulate expression of NDRG1 by directly targeting the NDRG1 3'-untranslated region. In conclusion, our results suggest that miR-182 plays an important role in the proliferation of human prostate cancer cells by directly suppressing the tumor supressor gene NDRG1. We uncovered a new epigenetic regulation of NDRG1.