EMAIL THIS PAGE TO A FRIEND
Terapevticheskii arkhiv
[Phosphate binders in chronic kidney disease: the positions of sevelamer].
PMID 23875200
Abstract
The paper shows the role of phosphate binders in the correction of phosphorus and calcium metabolic disturbances in chronic kidney disease. The results of clinical trials demonstrating the efficacy and safety of sevelamer are discussed.