EMAIL THIS PAGE TO A FRIEND
Communications in agricultural and applied biological sciences
The combined effects of DOC and salinity on the accumulation and toxicity of copper in mussel larvae.
PMID 23875291
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
BCR058
Continuous cast copper (O), BCR® certified Reference Material, rod
Cu
31284
Copper, electrolytic, Analytical Reagent, Reag. Ph. Eur., ≥99.9% (complexometric), wire
Cu
326445
Copper, bars (random sizes), ≥99.999% trace metals basis
Cu
254177
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Cu
326488
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Cu
266744
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Cu
349208
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Cu
12816
Copper, foil, ≥99.8% (complexometric)
Cu
203122
Copper, powder, 99.999% trace metals basis
Cu
266086
Copper, powder, <425 μm, 99.5% trace metals basis
Cu
207780
Copper, powder, <75 μm, 99%
Cu
326453
Copper, powder (spheroidal), 14-25 μm, 99%
Cu
357456
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Cu
365327
Copper, rod, diam. 11 mm, 99.9999% trace metals basis
Cu
223409
Copper, shot, −3-+14 mesh, 99%
Cu
520381
Copper, spherical shot, −20-+30 mesh, ≥99.5% trace metals basis
Cu
349224
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Cu
349216
Copper, wire, diam. 1.0 mm, ≥99.9%
Cu
326410
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Cu
326429
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Cu
61139
Copper, turnings, purum p.a., ≥99.0%
Cu
61141
Copper, purum p.a., ≥99.0%, powder
Cu
IRMM522A
Copper, IRMM® certified Reference Material, 0.1 mm foil
Cu
IRMM522B
Copper, IRMM® certified Reference Material, 1.0 mm foil
Cu
IRMM522C
Copper, IRMM® certified Reference Material, 0.5 mm wire
Cu
IRMM522D
Copper, IRMM® certified Reference Material, 1.0 mm wire
Cu
349151
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Cu
311405
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Cu
326437
Copper, rod, diam. 3.1 mm, 99.999%
Cu
767476
Copper, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Cu