EMAIL THIS PAGE TO A FRIEND
Communications in agricultural and applied biological sciences
Can prenatal nutritional constraints in the chicken embryo have long-term effects on the post-hatch performance and glucose metabolism?
PMID 23875301