EMAIL THIS PAGE TO A FRIEND
Leukemia research
Targeting hyaluronic acid production for the treatment of leukemia: treatment with 4-methylumbelliferone leads to induction of MAPK-mediated apoptosis in K562 leukemia.
PMID 23876826
Abstract
The current study examined the effect of modulation of hyaluronic acid (HA) synthesis on leukemia cell survival using the hyaluronic acid synthesis inhibitor 4-methylumbelliferone (4-MU). Treatment of CML cells with 4-MU led to caspase-dependent apoptosis characterized by decreased HA production, PARP cleavage, and increased phosphorylation of p38. Addition of exogenous HA, the pan caspase inhibitor Z-VAD-FMK or the p38 inhibitor SB203580 to 4-MU treated cells was able to protect cells from apoptosis. Treatment of tumor-bearing mice with 4-MU led to a significant reduction in tumor load which was mediated through the induction of apoptosis.