EMAIL THIS PAGE TO A FRIEND
Nanoscale
Multifunctional superparamagnetic iron oxide nanoparticles: design, synthesis and biomedical photonic applications.
PMID 23877222
Abstract
Superparamagnetic iron oxide nanoparticles (SPIONs) have shown great promise in biomedical applications. In this review, we summarize the recent advances in the design and fabrication of core-shell and hetero-structured SPIONs and further outline some exciting developments and progresses of these multifunctional SPIONs for diagnosis, multimodality imaging, therapy, and biophotonics.