EMAIL THIS PAGE TO A FRIEND
Archiv fur Kriminologie
[Suicide under the influence of "magic mushrooms"].
PMID 23878898
Abstract
Psilocybin/psilocin from so-called psychoactive mushrooms causes hallucinogenic effects. Especially for people with mental or psychiatric disorders ingestion of magic mushrooms may result in horror trips combined with the intention of self-destruction and suicidal thoughts. Automutilation after consumption of hallucinogenic mushrooms has already been described. Our case report demonstrates the suicide of a man by self-inflicted cut and stab injuries. A causal connection between suicidal behaviour and previous ingestion of psychoactive mushrooms is discussed.