EMAIL THIS PAGE TO A FRIEND
Cancer letters
Synergistic effect of combined treatment with gamma-tocotrienol and statin on human malignant mesothelioma cells.
PMID 23879968
Abstract
The present study is the first to demonstrate the synergetic effect of statins (atorvastatin and simvastatin) and gamma-tocotrienol (γ-T3) on human malignant mesothelioma (MM). Statin + γ-T3 combinations induced greater cell growth inhibition more than each single treatment via inhibition of mevalonate pathway, a well-known target of both γ-T3 and statins. γ-T3 was necessary for endoplasmic reticulum stress markers CHOP and GRP78, whereas an intrinsic apoptotic marker, caspase 3 activation was induced only in the presence of statins. Overall, the combination of γ-T3 and statins could be useful for MM therapy and functions in a complementary style.