EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
Metabolic engineering strategies for improving xylitol production from hemicellulosic sugars.
PMID 23881318
Abstract
Xylitol is a five-carbon sugar alcohol with potential for use as a sweetener. Industrially, xylitol is currently produced by chemical hydrogenation of D-xylose using Raney nickel catalysts and this requires expensive separation and purification steps as well as high pressure and temperature that lead to environmental pollution. Highly efficient biotechnological production of xylitol using microorganisms is gaining more attention and has been proposed as an alternative process. Although the biotechnological method has not yet surpassed the advantages of chemical reduction in terms of yield and cost, various strategies offer promise for the biotechnological production of xylitol. In this review, the focus is on the most recent developments of the main metabolic engineering strategies for improving the production of xylitol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
X0080000
Xylitol, European Pharmacopoeia (EP) Reference Standard
C5H12O5