EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
Anodic TiO2 nanotubular arrays with pre-synthesized hydroxyapatite--an effective approach to enhance the biocompatibility of titanium.
PMID 23882759
Abstract
Electrochemically anodized TiO2 nanotubular arrays can provide large surface areas for biological species attachment. In order to further enhance the biocompatibility of Ti medical implants, we deposited a pre-synthesized hydroxyapatite inside and on the nanotubular arrays, and examined the biocompatibility of the anodized TiO2 nanotubular arrays with pre-synthesized hydroxyapatite by in vitro assessment in simulated body fluid, and in vitro cell culture. The results showed that the hydroxyapatite coating was able to be induced on TiO2 nanotubular arrays with pre-synthesized hydroxyapatite within 5 days while only a thin film composed of calcium phosphorous chemicals formed on as-formed TiO2 nanotubular arrays. The cell culture evaluation further proved the enhancement of cell attachment and proliferation on TiO2 nanotubular arrays with pre-synthesized hydroxyapatite as opposed to those without pre-synthesized hydroxyapatite. The present study proves that formation of TiO2 nanotubular arrays with pre-synthesized hydroxyapatite a promising method to enhance the biocompatibility of Ti implants.