EMAIL THIS PAGE TO A FRIEND
Revue medicale suisse
[Endometriosis, an underestimated disease: reducing pelvic pain effectively and lastingly].
PMID 23882918
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7252
19-Nortestosterone
C18H26O2