EMAIL THIS PAGE TO A FRIEND
Therapeutic delivery
Recent advances in porous silicon technology for drug delivery.
PMID 23883125
Abstract
Porous silicon (pSi) is a nanostructured carrier system that has received considerable attention over the past 10 years, for use in a wide variety of biomedical applications, including biosensing, biomedical imaging, tissue scaffolds and drug delivery. This interest is due to several key features of pSi, including excellent in vivo biocompatibility, the ease of surface chemistry modification and the control over its 3D porous network structure. With control of these physical parameters pSi has successfully been used for the delivery of a variety of therapeutics, ranging from small-molecule drugs to larger peptide/protein-type therapeutics. In this review, the authors provide a brief overview of pSi fabrication methods, particularly with regard to the need to passivate the highly reactive Si-Hx surface species of native pSi, typically via thermal oxidation, hydrocarbonization or hydrosilylation. This surface modification, in turn, controls both the loading and release of therapeutics. The authors will then report on specific case studies of leading examples on the use of pSi as a therapeutic-delivery system. Specifically, the first reported in vivo study that demonstrated the use of pSi to improve the delivery of a Biopharmaceutical Classification System Class 2 poorly soluble drug (indomethacin), by using thermally oxidized pSi, is discussed, as well as highlighting a study that determined the biodistribution of (18)F-radiolabeled thermally hydrocarbonized pSi after oral dosing. The authors also report on the development of composite pSi-poly(D,L-lactide-co-glycolide) microparticles for the controlled delivery of protein therapeutics. Finally, the use of pSi in the delivery of bioactives, such as the successful use of thermally carbonized pSi to deliver Melanotan II, an unspecific agonist for the melanocortin receptors that are involved in controlling fluid uptake is discussed. With a growing body of literature reporting the successful use of pSi to deliver a range of therapeutics, we are entering what may be a golden age for this drug-delivery system, which may finally see the long-held promises finally achieved.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
343250
Silicon, pieces, 99.95% trace metals basis
Si
267414
Silicon, powder, −60 mesh, 99.999% trace metals basis
Si
215619
Silicon, powder, −325 mesh, 99% trace metals basis
Si
633097
Silicon, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis
Si
646687
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Si
647535
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Si
647780
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Si
647802
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 3 in. × 0.5 mm
Si
647675
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm
Si
647764
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Si
647101
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Si
647543
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Si
647799
Silicon, wafer (single side polished), contains phosphorus as dopant, <111>, N-type, diam. × thickness 2 in. × 0.5 mm
Si
647705
Silicon, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Si
647772
Silicon, wafer (single side polished), <111>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Si
767492
Silicon, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.999% trace metals basis
Si
GF37993595
Silicon, rod, 100mm, diameter 10.0mm, single crystal - random orientation, 100%
Si
GF64686998
Silicon, rod, 100mm, diameter 2.0mm, crystalline, 100%
Si
GF22929307
Silicon, rod, 100mm, diameter 5.0mm, crystalline, 100%
Si
GF16363391
Silicon, rod, 10mm, diameter 2.0mm, crystalline, 100%
Si
GF66465276
Silicon, rod, 25mm, diameter 2.0mm, crystalline, 100%
Si
GF43555664
Silicon, rod, 25mm, diameter 3.15mm, single crystal - random orientation, 100%
Si
GF70438518
Silicon, rod, 40mm, diameter 20mm, single crystal, -100, 99.999%
Si
GF35274113
Silicon, rod, 40mm, diameter 20mm, single crystal, -111, 99.999%
Si
GF05482852
Silicon, rod, 50mm, diameter 2.0mm, crystalline, 100%
Si
GF91515034
Silicon, rod, 50mm, diameter 3.15mm, single crystal - random orientation, 100%
Si
GF14076930
Silicon, rod, 50mm, diameter 5.0mm, crystalline, 100%
Si
GF28448691
Silicon, rod, 50mm, diameter 5mm, single crystal, -100, 99.999%
Si
GF40945754
Silicon, rod, 50mm, diameter 5mm, single crystal, -111, 99.999%
Si
GF64342188
Silicon, rod, 50mm, diameter 6mm, single crystal, -111, 99.999%
Si